Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aebp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178803.2 | NP_848918.1 | 5 | 950 | ||
GenBank mRNA | AB221647.1 | - | 5 | 601 | ||
AF090326.1 | - | 685 | ||||
AK029011.1 | - | 5 | 952 | |||
AK045838.1 | - | 5 | 716 | |||
AK045987.1 | - | 5 | 716 | |||
AK155272.1 | - | 5 | 682 | |||
BC025467.1 | - | 5 | 1408 | 5351959 | ||
BC031376.1 | - | 6 | 811 | 5356616 | ||
BC057681.1 | - | 5 | 730 | 3499299 |