Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Boc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172506.2 | NP_766094.1 | 2, 3 | 598 | ||
GenBank mRNA | AF388037.1 | - | 1, 2 | 51 | ||
AK028770.1 | - | 2, 3 | 619 | |||
AK028889.1 | - | 2, 3 | 599 | |||
AK043379.1 | - | 2, 3 | 608 | |||
BC062892.1 | - | 2, 3 | 578 | 6418202 |