Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arntl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243048.1 | NP_001229977.1 | 10 | 1130 | ||
NM_007489.4 | NP_031515.1 | 13 | 1423 | |||
GenBank mRNA | AB012601.1 | - | 10 | 921 | ||
AB012602.1 | - | 9 | 875 | |||
AB014494.1 | - | 13 | 1309 | |||
AB015203.1 | - | 9 | 900 | |||
AK146628.1 | - | 13 | 1423 | |||
AK147145.1 | - | 13 | 1423 | |||
BC011080.1 | - | 10 | 1081 | 3661605 | ||
BC025973.1 | - | 11 | 1174 | 3980719 |