Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cpox | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007757.2 | NP_031783.2 | 4, 5 | 1150 | ||
GenBank mRNA | AK089471.1 | - | 1 | 1116 | ||
AK157277.1 | - | 4, 5 | 1033 | |||
AK166512.1 | - | 4, 5 | 1013 | |||
BC017680.1 | - | 4, 5 | 1007 | 4241687 | ||
CA535096.1 | - | 4, 5 | 478 | |||
D16333.1 | - | 4, 5 | 1064 |