Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eps8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271587.1 | NP_001258516.1 | 14 | 1586 | ||
NM_001271588.1 | NP_001258517.1 | 14 | 1517 | |||
NM_001271589.1 | NP_001258518.1 | 7 | 587 | |||
NM_001271595.1 | NP_001258524.1 | 14 | 1591 | |||
NM_007945.3 | NP_031971.2 | 15 | 1592 | |||
GenBank mRNA | AK136246.1 | - | 5 | 358 | ||
AK147877.1 | - | 15 | 1520 | |||
AK148424.1 | - | 15 | 1508 | |||
AK149683.1 | - | 14 | 1380 | |||
AK155022.1 | - | 14 | 1352 | |||
AK165684.1 | - | 7 | 590 | |||
AK166156.1 | - | 15 | 1427 | |||
AK187916.1 | - | 2 | 64 | |||
BC016890.1 | - | 14 | 1603 | 4240899 | ||
L21671.1 | - | 14 | 1507 |