Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019395.3 | NP_062268.1 | 2, 3 | 583 | ||
GenBank mRNA | AJ132693.1 | - | 2, 3 | 534 | ||
AK002216.1 | - | 2, 3 | 445 | |||
AK149527.1 | - | 2, 3 | 464 | |||
BC011480.1 | - | 2, 3 | 447 | 4194764 | ||
BC051392.1 | - | 2, 3 | 447 | 5054854 |