Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fkbp5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010220.4 | NP_034350.1 | 3 | 368 | ||
GenBank mRNA | AK169795.1 | - | 3 | 309 | ||
BC015260.1 | - | 3 | 307 | 4237766 | ||
BU613862.1 | - | 1 | 35 | |||
BY740772.1 | - | 3 | 368 | |||
CA317455.1 | - | 3 | 331 | |||
CT010372.1 | - | 2 | 149 | |||
U16959.1 | - | 3 | 368 | |||
U36220.1 | - | 3 | 296 |