Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fzd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021458.2 | NP_067433.1 | 4 | 1166 | ||
GenBank mRNA | AK134261.1 | - | 4 | 1102 | ||
BC050965.1 | - | 4 | 1127 | |||
BC066186.1 | - | 4 | 826 | |||
BC103714.1 | - | 4 | 826 | |||
U43205.1 | - | 3 | 1015 |
Set Membership: |
![]() |