Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gna12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010302.2 | NP_034432.1 | 4 | 766 | ||
GenBank mRNA | AK194048.1 | - | 3 | 164 | ||
BC111810.1 | - | 5 | 533 | 40045663 | ||
M63659.1 | - | 4 | 766 |