Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Idua | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008325.4 | NP_032351.2 | 5 | 845 | ||
NM_172997.1 | NP_766585.1 | 4 | 704 | |||
GenBank mRNA | AK031414.1 | - | 4 | 499 | ||
AK045330.1 | - | 3 | 332 | |||
AK155627.1 | - | 5 | 846 | |||
AK169998.1 | - | 4 | 503 | |||
BC057935.1 | - | 4 | 466 | 5327150 | ||
L34111.1 | - | 4 | 419 |