Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Igf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001122736.1 | NP_001116208.1 | 2, 3 | 265 | ||
NM_001122737.1 | NP_001116209.1 | 2, 3 | 255 | |||
NM_010514.3 | NP_034644.2 | 2, 3 | 602 | |||
GenBank mRNA | BC053489.1 | - | 2, 3 | 241 | 30013295 | |
BC058615.1 | - | 2, 3 | 240 | 5686066 | ||
CF742200.1 | - | 2, 3 | 461 | |||
CN460564.1 | - | 2, 3 | 477 | |||
M14951.1 | - | 2, 3 | 186 |