Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Itgam | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001082960.1 | NP_001076429.1 | 17 | 2221 | ||
NM_008401.2 | NP_032427.2 | 17 | 2218 | |||
GenBank mRNA | AK040921.1 | - | 7 | 947 | ||
AK089521.1 | - | 17 | 2185 | |||
AK146569.1 | - | 9 | 1123 | |||
AK154224.1 | - | 17 | 2138 | |||
AK154474.1 | - | 17 | 2172 | |||
AK154798.1 | - | 17 | 2134 | |||
AK155006.1 | - | 17 | 2150 | |||
AK155520.1 | - | 17 | 2143 | |||
AK155671.1 | - | 17 | 2187 | |||
AK155717.1 | - | 17 | 2176 | |||
AK170324.1 | - | 17 | 2190 | |||
AK171068.1 | - | 14 | 1850 | |||
EF101557.1 | - | 3 | 416 |