Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Glmn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161738.1 | NP_001155210.1 | 11 | 1142 | ||
NM_001161739.1 | NP_001155211.1 | 11 | 1156 | |||
NM_133248.2 | NP_573511.2 | 11 | 1156 | |||
GenBank mRNA | AJ566083.1 | - | 10 | 1024 | ||
AK034160.1 | - | 11 | 1143 | |||
AK165661.1 | - | 11 | 1127 | |||
AK169045.1 | - | 11 | 1128 | |||
AK172659.1 | - | 11 | 1094 | |||
BC003446.1 | - | 11 | 1092 | 2651614 |