Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mgrn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252437.1 | NP_001239366.1 | 9, 10 | 1082 | ||
NM_029657.4 | NP_083933.1 | 9, 10 | 1079 | |||
GenBank mRNA | AK011747.2 | - | 9, 10 | 872 | ||
AK088533.1 | - | 9, 10 | 862 | |||
AK129161.1 | - | 6, 7 | 490 | |||
AK150176.1 | - | 10, 11 | 915 | |||
AK153407.1 | - | 9, 10 | 903 | |||
AK186778.1 | - | 1, 2 | 61 | |||
BC046830.1 | - | 9, 10 | 928 | 6811606 |