Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mrvi1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010826.5 | NP_034956.2 | 8, 9 | 1498 | ||
NM_194464.3 | NP_919446.2 | 9, 10 | 1741 | |||
GenBank mRNA | AK141821.1 | - | 4, 5 | 780 | ||
AK157169.1 | - | 9, 10 | 1351 | |||
U63407.1 | - | 9, 10 | 1744 | |||
U63408.1 | - | 8, 9 | 1493 |