Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010897.2 | NP_035027.1 | 44, 45 | 6821 | ||
GenBank mRNA | AK033306.1 | - | 4, 5 | 665 | ||
AK158728.1 | - | 7, 8 | 1326 | |||
L10370.1 | - | 43, 44 | 6666 | |||
X54924.1 | - | 5, 6 | 866 |