Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: P2ry1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282016.1 | NP_001268945.1 | 1 | 1701 | ||
NM_008772.5 | NP_032798.1 | 1 | 1701 | |||
GenBank mRNA | AK030759.1 | - | 1 | 1690 | ||
AK033660.1 | - | 1 | 1687 | |||
AK036611.1 | - | 1 | 1692 | |||
BC138634.1 | - | 1 | 1562 | 8861654 | ||
BC138635.1 | - | 1 | 1562 | 8861655 | ||
EF584663.1 | - | 1 | 53 | |||
U22829.1 | - | 1 | 1075 |
Set Membership: |
G-protein coupled receptor |