Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prkca | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011101.3 | NP_035231.2 | 16 | 1997 | ||
GenBank mRNA | AK147084.1 | - | 16 | 1998 | ||
AK163950.1 | - | 6 | 685 | |||
BC096493.1 | - | 16 | 1791 | 30742571 | ||
M25811.1 | - | 16 | 2031 | |||
X52684.1 | - | 17 | 1938 | |||
X52685.1 | - | 17 | 1938 |