Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Npepps | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008942.2 | NP_032968.2 | 6 | 791 | ||
GenBank mRNA | AK133898.1 | - | 6 | 599 | ||
BC009653.1 | - | 6 | 730 | 3487224 | ||
BC086798.1 | - | 6 | 783 | 6812900 | ||
BC098212.1 | - | 6 | 791 | 6825181 | ||
U35646.1 | - | 6 | 778 |