Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptdss1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008959.3 | NP_032985.2 | 12, 13 | 1986 | ||
GenBank mRNA | AF042731.1 | - | 12, 13 | 1401 | ||
AK036990.1 | - | 12, 13 | 1448 | |||
AK054101.1 | - | 12, 13 | 1425 | |||
AK085857.1 | - | 12, 13 | 1987 | |||
AK086136.1 | - | 12, 13 | 1645 | |||
AK088048.1 | - | 12, 13 | 1468 | |||
AK137681.1 | - | 12, 13 | 1656 | |||
AK172873.1 | - | 11, 12 | 3772 | |||
AK219321.1 | - | 1, 2 | 75 | |||
BC003260.1 | - | 12, 13 | 1623 | 3501141 |