Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rlim | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011276.3 | NP_035406.3 | 5 | 1341 | ||
GenBank mRNA | AF069992.1 | - | 5 | 1296 | ||
AK013207.1 | - | 5 | 1313 | |||
AK029295.1 | - | 5 | 1335 | |||
BC012960.1 | - | 3 | 1074 | 4193003 |