Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnf43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172448.3 | NP_766036.2 | 9 | 2889 | ||
GenBank mRNA | AK028750.1 | - | 8 | 2248 | ||
BC029717.1 | - | 1 | 1265 | |||
BC037465.1 | - | 1 | 1329 | |||
BC131957.1 | - | 7 | 2007 | 40130234 |