Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Srpk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016795.3 | NP_058075.2 | 14 | 1698 | ||
GenBank mRNA | AB012290.1 | - | 13 | 1656 | ||
AJ224115.1 | - | 14 | 1692 | |||
AK015754.1 | - | 10 | 1226 | |||
AK140761.1 | - | 1108 | ||||
AK151152.1 | - | 14 | 1761 | |||
BC005707.1 | - | 14 | 1667 | 3157835 | ||
BC050761.1 | - | 14 | 1698 | 6771151 |