Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mylk3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001297612.1 | NP_001284541.1 | 7 | 1338 | ||
NM_175441.5 | NP_780650.2 | 7 | 1791 | |||
GenBank mRNA | AK052858.1 | - | 7 | 1626 | ||
AK146683.1 | - | 7 | 1792 | |||
AK165216.1 | - | 7 | 1280 | |||
BC137983.1 | - | 7 | 1764 | 8861002 | ||
BC145079.1 | - | 7 | 1765 | |||
EU403565.1 | - | 7 | 1734 |