Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Senp6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146003.2 | NP_666115.2 | 23 | 3841 | ||
GenBank mRNA | AK028461.1 | - | 11 | 1525 | ||
AK048485.1 | - | 18 | 3290 | |||
BC052718.1 | - | 23 | 3608 |