Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tgm2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009373.3 | NP_033399.1 | 9 | 1242 | ||
GenBank mRNA | AF076928.1 | - | 8 | 1157 | ||
AF114266.1 | - | 9 | 1207 | |||
AK052912.1 | - | 9 | 1217 | |||
AK080224.1 | - | 9 | 1242 | |||
AK080593.1 | - | 9 | 1217 | |||
AK086001.1 | - | 9 | 1218 | |||
AK089481.1 | - | 9 | 1245 | |||
AK143712.1 | - | 9 | 1278 | |||
AK151776.1 | - | 9 | 1243 | |||
AK152152.1 | - | 9 | 1217 | |||
AK152627.1 | - | 9 | 1243 | |||
AK159255.1 | - | 9 | 1217 | |||
AK166302.1 | - | 9 | 1217 | |||
AK168990.1 | - | 9 | 1218 | |||
AK169356.1 | - | 9 | 1245 | |||
BC016492.1 | - | 9 | 1183 | 3256943 | ||
M55154.1 | - | 9 | 1179 |