Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Umod | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278605.1 | NP_001265534.1 | 8, 9 | 1897 | ||
NM_009470.5 | NP_033496.1 | 8, 9 | 1953 | |||
GenBank mRNA | AK085460.1 | - | 8, 9 | 1947 | ||
AK144065.1 | - | 8, 9 | 1869 | |||
AK164688.1 | - | 8, 9 | 1869 | |||
AK165507.1 | - | 8, 9 | 1813 | |||
BC012973.1 | - | 7, 8 | 1791 | 4210940 |