Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Erlin1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164359.1 | NP_001157831.1 | 10 | 897 | ||
NM_001164360.1 | NP_001157832.1 | 10 | 805 | |||
NM_145502.3 | NP_663477.3 | 9 | 1001 | |||
GenBank mRNA | AK133557.1 | - | 9 | 744 | ||
AK145990.1 | - | 10 | 778 | |||
BC011220.1 | - | 10 | 769 | 4208238 | ||
BC023849.1 | - | 9 | 864 | 5345801 |