Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ikbkap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026079.3 | NP_080355.2 | 25 | 2880 | ||
GenBank mRNA | AF140786.1 | - | 25 | 2808 | ||
AF367244.1 | - | 25 | 2833 | |||
AF387811.1 | - | 25 | 2847 | |||
AK031367.1 | - | 25 | 2825 | |||
AK035944.1 | - | 25 | 2882 | |||
AK147153.1 | - | 25 | 2883 | |||
AK165168.1 | - | 22 | 2357 | |||
BC052387.1 | - | 25 | 2792 | 5705589 |