Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gm15455 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161816.1 | NP_001155288.1 | 1 | 2767 | ||
GenBank mRNA | AK139531.1 | - | 1538 | |||
BC157984.1 | - | 1 | 2691 | |||
BC157993.1 | - | 1 | 2691 |
Gene Symbol: Rbm10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167775.1 | NP_001161247.1 | 24 | 3468 | ||
NM_001167776.1 | NP_001161248.1 | 23 | 2953 | |||
NM_145627.2 | NP_663602.1 | 24 | 3184 | |||
GenBank mRNA | AK089105.1 | - | 23 | 2764 | ||
AK122208.1 | - | 21 | 4884 | |||
AK145991.1 | - | 24 | 3184 | |||
AK153736.1 | - | 24 | 3469 | |||
AK167666.1 | - | 23 | 2931 | |||
BC004674.1 | - | 23 | 2869 | 3500403 |