Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gnl3l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168600.1 | NP_001162071.1 | 14 | 1462 | ||
NM_198110.2 | NP_932778.1 | 14 | 1495 | |||
GenBank mRNA | BC020354.1 | - | 14 | 1431 | ||
BC057033.1 | - | 14 | 1421 | 6834283 | ||
BC079653.1 | - | 14 | 1417 | 6843880 |