Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Npc1l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207242.2 | NP_997125.2 | 6 | 2213 | ||
GenBank mRNA | AK078947.1 | - | 6 | 2114 | ||
AY437866.1 | - | 6 | 2091 | |||
BC131788.1 | - | 6 | 2090 | |||
BC131789.1 | - | 6 | 2091 | 40086103 |