Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Papss1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289477.1 | NP_001276406.1 | 12, 13 | 1888 | ||
NM_001289478.1 | NP_001276407.1 | 12, 13 | 2075 | |||
NM_001289479.1 | NP_001276408.1 | 13, 14 | 2186 | |||
NM_011863.2 | NP_035993.1 | 11, 12 | 1898 | |||
GenBank mRNA | AK146507.1 | - | 11, 12 | 1771 | ||
AK153296.1 | - | 11, 12 | 1768 | |||
AK160875.1 | - | 13, 14 | 2090 | |||
AK168982.1 | - | 11, 12 | 1771 | |||
AK169117.1 | - | 12, 13 | 1950 | |||
AK169124.1 | - | 11, 12 | 1765 | |||
AK169204.1 | - | 12, 13 | 1950 | |||
BC066055.1 | - | 12, 13 | 1954 | 6833756 | ||
U34883.1 | - | 11, 12 | 1775 |