Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fem1c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173423.4 | NP_775599.1 | 3 | 1951 | ||
GenBank mRNA | AK028330.1 | - | 3 | 1952 | ||
AK049336.1 | - | 3 | 1939 | |||
AK129440.1 | - | 1884 | ||||
AK147729.1 | - | 3 | 1841 | |||
AY249189.1 | - | 2 | 1413 | |||
BC138604.1 | - | 2 | 1494 | 8861624 | ||
BC138605.1 | - | 2 | 1494 | 8861625 |