Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klhl9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172871.2 | NP_766459.2 | 1 | 600 | ||
GenBank mRNA | AK033231.1 | - | 1 | 558 | ||
AK049803.1 | - | 1 | 555 | |||
AK129338.1 | - | 1 | 502 | |||
BC094556.1 | - | 1 | 542 | 6811241 |