Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppp2r5c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081457.2 | NP_001074926.1 | 6, 7 | 805 | ||
NM_001081458.2 | NP_001074927.1 | 6, 7 | 805 | |||
NM_001135001.1 | NP_001128473.1 | 8, 9 | 1036 | |||
NM_012023.3 | NP_036153.2 | 6, 7 | 805 | |||
GenBank mRNA | AB055635.1 | - | 6, 7 | 655 | ||
AB055636.1 | - | 6, 7 | 655 | |||
AK031584.1 | - | 6, 7 | 808 | |||
AK129042.1 | - | 8, 9 | 908 | |||
AK191098.1 | - | 1, 2 | 21 | |||
AK196453.1 | - | 1, 2 | 21 | |||
BC003979.1 | - | 6, 7 | 765 | 3489999 | ||
U37353.1 | - | 6, 7 | 634 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AK012612.1 | - | 920 | Chr. - - [-] on Build |