Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppp2r5c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081457.2 | NP_001074926.1 | 4 | 566 | ||
NM_001081458.2 | NP_001074927.1 | 4 | 566 | |||
NM_001135001.1 | NP_001128473.1 | 6 | 797 | |||
NM_012023.3 | NP_036153.2 | 4 | 566 | |||
GenBank mRNA | AB055635.1 | - | 4 | 416 | ||
AB055636.1 | - | 4 | 416 | |||
AK031584.1 | - | 4 | 569 | |||
AK129042.1 | - | 6 | 669 | |||
BC003979.1 | - | 4 | 526 | 3489999 | ||
CV558022.1 | - | 4 | 566 | |||
U37353.1 | - | 4 | 395 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AK012612.1 | - | 1159 | Chr. - - [-] on Build |