Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sestd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175465.6 | NP_780674.1 | 8, 9 | 699 | ||
GenBank mRNA | AK047939.1 | - | 8, 9 | 697 | ||
BC013245.1 | - | 9, 10 | 780 | |||
BC044853.1 | - | 8, 9 | 699 | 3978465 | ||
BG082832.2 | - | 5, 6 | 442 | |||
CO425092.1 | - | 2, 3 | 132 |