Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177779.3 | NP_808447.2 | 6, 7 | 1050 | ||
GenBank mRNA | AK079909.1 | - | 2, 3 | 479 | ||
BC138437.1 | - | 2, 3 | 477 | 8861457 | ||
BC138438.1 | - | 2, 3 | 477 | 8861458 |