Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tnpo3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177296.4 | NP_796270.2 | 7, 8 | 1135 | ||
GenBank mRNA | AK145723.1 | - | 7, 8 | 1152 | ||
BC064646.1 | - | 7, 8 | 1135 | 6313452 | ||
BC075678.1 | - | 7, 8 | 1348 | 30619729 |