Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Efr3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001082483.1 | NP_001075952.1 | 5 | 628 | ||
GenBank mRNA | AK129247.1 | - | 1 | 75 | ||
BC075650.1 | - | 4 | 385 | |||
BC147643.1 | - | 5 | 628 | 9056633 | ||
BC157968.1 | - | 5 | 628 | 9088136 |