Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kif14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001287179.2 | NP_001274108.1 | 1 | 676 | ||
GenBank mRNA | AB731728.1 | - | 1 | 566 | ||
AK087451.1 | - | 2 | 705 | |||
BC158132.1 | - | 1 | 589 | 9007399 |