Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scn4b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013390.2 | NP_001013408.1 | 2, 3 | 228 | ||
GenBank mRNA | AB118946.1 | - | 2, 3 | 515 | ||
AY669079.1 | - | 2, 3 | 228 | |||
BC139399.1 | - | 2, 3 | 307 | 8862421 | ||
BC139400.1 | - | 2, 3 | 307 | 8862422 |