Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rufy4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034060.3 | NP_001029232.2 | 6 | 614 | ||
NM_001170641.1 | NP_001164112.1 | 6 | 637 | |||
GenBank mRNA | AK158260.1 | - | 6 | 619 | ||
BC150907.1 | - | 6 | 605 | 9087818 | ||
BC150910.1 | - | 6 | 605 | 9087821 |