Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam92b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033980.2 | NP_001029152.2 | 3 | 436 | ||
GenBank mRNA | AK131790.1 | - | 3 | 436 | ||
BC147233.1 | - | 3 | 355 | 9056283 | ||
BC147234.1 | - | 3 | 355 | 9056269 | ||
BC172014.1 | - | 2 | 120 |