Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc25a13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177572.1 | NP_001171043.1 | 18 | 2031 | ||
NM_015829.3 | NP_056644.1 | 18 | 2034 | |||
GenBank mRNA | AF164632.1 | - | 18 | 1979 | ||
AK002829.1 | - | 18 | 1909 | |||
AK012670.1 | - | 18 | 1922 | |||
AK029020.1 | - | 18 | 1962 | |||
AK150496.1 | - | 18 | 1976 | |||
AK151180.1 | - | 18 | 1976 | |||
AK152237.1 | - | 18 | 1976 | |||
BC016571.1 | - | 12 | 1168 | 3600909 |