Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zwint | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001293683.1 | NP_001280612.1 | 1, 2 | 122 | ||
NM_001293684.1 | NP_001280613.1 | 1, 2 | 122 | |||
NM_025635.4 | NP_079911.1 | 1, 2 | 122 | |||
GenBank mRNA | AK004305.1 | - | 1, 2 | 74 | ||
AK008144.1 | - | 1, 2 | 72 | |||
AK013556.1 | - | 1, 2 | 57 | |||
AK075977.1 | - | 1, 2 | 68 | |||
AK082215.1 | - | 1, 2 | 72 | |||
AK134380.1 | - | 1, 2 | 74 | |||
AK184469.1 | - | 1, 2 | 21 | |||
BC013559.1 | - | 1, 2 | 40 | |||
BC027100.1 | - | 1, 2 | 79 | 5359154 | ||
BC034870.1 | - | 1, 2 | 51 | 1382192 | ||
BY240779.1 | - | 1, 2 | 123 |