Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rabep1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291141.1 | NP_001278070.1 | 11 | 1954 | ||
NM_001291142.1 | NP_001278071.1 | 11 | 1963 | |||
NM_001291143.1 | NP_001278072.1 | 13 | 2154 | |||
NM_019400.3 | NP_062273.2 | 12 | 2083 | |||
GenBank mRNA | AB001750.1 | - | 13 | 1990 | ||
AF248489.1 | - | 11 | 1749 | |||
AF248490.1 | - | 11 | 1740 | |||
AK134587.1 | - | 12 | 2084 | |||
BC003921.1 | - | 3 | 704 | |||
BC060166.1 | - | 12 | 2101 | |||
BC080274.1 | - | 12 | 2055 | |||
BC129853.1 | - | 12 | 1902 | 40109688 | ||
BC129854.1 | - | 12 | 1902 | 40109689 | ||
D86066.1 | - | 12 | 1919 |