Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Akap8l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017476.2 | NP_059504.2 | 5 | 458 | ||
GenBank mRNA | AB028921.1 | - | 5 | 451 | ||
AK011325.1 | - | 5 | 463 | |||
AK166526.1 | - | 6 | 374 | |||
AK195897.1 | - | 2 | 215 | |||
BC046801.1 | - | 5 | 447 | 6403966 | ||
BC051015.1 | - | 5 | 458 |