Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Paxip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018878.3 | NP_061366.2 | 11 | 2404 | ||
GenBank mRNA | AF104261.1 | - | 11 | 2404 | ||
AK144606.1 | - | 11 | 2266 | |||
AK148141.1 | - | 11 | 2274 | |||
BC066014.1 | - | 11 | 2193 | 5704566 |